Learn More
R&D Systems™ Mouse CD7 (NP_033984) VersaClone cDNA
Description
- CD7 is a transmembrane protein which is a member of the immunoglobulin superfamily
- It is found on thymocytes and mature Tcells and plays an essential role in Tcell interactions and also in T-cell/Bcell interaction during early lymphoid development
- CD7 is a negative prognostic marker in myeloid malignancies
- The CD7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mCd7 |
| Accession Number | NP_033984 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 646 bp |
| Gene ID (Entrez) | 924 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |