missing translation for 'onlineSavingsMsg'
Learn More

Molybdatophosphoric Acid Hydrate, Supelco™

Product Code. 15813910
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15813910 100 g 100g
15803910 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15813910 Supplier Supelco Supplier No. 1005320100
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51429-74-4

Specifications

Melting Point 78°C to 90°C (anhydrous substance)
Density 1.62 g/cm3
Quantity 100 g
Molecular Formula H3[P(Mo3O10)4] · X H2O
MDL Number MFCD00149913
Synonym Molybdatophosphoric acid hydrate, Phosphomolybdic acid, Molybdatophosphoric acid hydrate
Molecular Weight (g/mol) 1825.25 (anhydrous basis)
Grade Ungraded
Chemical Name or Material Molybdatophosphoric Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.