accessibility menu, dialog, popup

UserName

A(a)A(a)

CAS RN 51429-74-4

IUPAC Name: phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
Synonyms: phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
Molecular Weight (g/mol): 1825.4
Molecular Formula: H3Mo12O40P
InChi Key: FEFSFHGZSNHJEL-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (18)
Quantity 
  • (1)
  • (8)
  • (7)
  • (3)
  • (4)
Grade 
  • (6)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (7)
  • (3)
  • (3)
  • (2)
  • (18)
  • (1)
  • (8)
  • (7)
  • (3)
  • (4)
  • (6)
  • (5)
  • (2)
  • (1)