missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mmp9 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | Mmp9 |
|---|---|
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18028775
|
Proteintech
30592-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18027315
|
Proteintech
30592-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mmp9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse, human antigen. The Mmp9 antibody has been validated for the following applications: WB,IF,ELISA analysis.
Specifications
| Mmp9 | |
| Western Blot, Immunofluorescence, ELISA | |
| Rabbit | |
| 17395 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 500 μg/mL | |
| Liquid | |
| Mouse, Human | |
| AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title