Learn More
Mmp9 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30592-1-AP-20UL
 View more versions of this product
Product Code. 18028775
  • 145.00 EUR / 20µL
Description
The Mmp9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse, human antigen. The Mmp9 antibody has been validated for the following applications: WB,IF,ELISA analysis.
Specifications
Specifications
Mmp9
500 μg/mL
Rabbit
Primary
Mouse, Human
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9
20 μL
17395
Store at -20°C. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions