Learn More
Mmp9 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30592-1-AP-150UL
This item is not returnable.
View return policy
Description
The Mmp9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse, human antigen. The Mmp9 antibody has been validated for the following applications: WB,IF,ELISA analysis.
Specifications
| Mmp9 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9 | |
| 150 μL | |
| 17395 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |