Learn More
Tocris Bioscience™ ML 162
Description
ML 162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. It induces ferroptosis in vivo. It is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50 = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).
Specifications
Specifications
| Chemical Name or Material | 2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide |
| CAS | 1035072-16-2 |
| Quantity | 10 mg |
| Target | Glutathione Peroxidase 4/GPX4 |
| Molecular Formula | C23H22Cl2N2O3S |
| Purity | 0.98 |