missing translation for 'onlineSavingsMsg'
Learn More

MK-2206 Dihydrochloride, TRC

Product Code. 30476188
Change view
Click to view available options
Quantity:
1 mg
25 mg
50 mg
Unit Size:
1mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476188 1 mg 1mg
30470196 50 mg 50mg
30468904 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476188 Supplier Toronto Research Chemicals Supplier No. TRCM4250251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1032350-13-2
Molecular Formula C25 H21 N5 O . 2 Cl H
Molecular Weight (g/mol) 480.39
Synonym MK 2206 dihydrochloride
IUPAC Name 8-[4-(1-aminocyclobutyl)phenyl]-9-phenyl-2H-[1,2,4]triazolo[3,4-f][1,6]naphthyridin-3-one;dihydrochloride
SMILES Cl.Cl.NC1(CCC1)c2ccc(cc2)c3nc4C=CN5C(=O)NN=C5c4cc3c6ccccc6

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C25H21N5O.2ClH/c26-25(12-4-13-25)18-9-7-17(8-10-18)22-19(16-5-2-1-3-6-16)15-20-21(27-22)11-14-30-23(20)28-29-24(30)31;;/h1-3,5-11,14-15H,4,12-13,26H2,(H,29,31);2*1H
Formula Weight 479.128
Chemical Name or Material MK-2206 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.