missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Storage is content dependent. |
| Quantity | 1 Kit |
| Kit Contents | 5X Homogenization Buffer 50mL, 5X Suspension Buffer 2 x 50mL, Mitochondrial Staining Solution 1mL, Mitochondria Lysis Buffer 50mL, 10X PBS 2 x 50mL, 1M DTT 100μL, 100X Protease Inhibitor Cocktail (PIC) 100μL |
| For Use With (Application) | Western Blotting |
| Product Type | Mitochondrial Extraction Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?