Learn More
Methyl Palmitoleate, TRC
Product Code. 30134268
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
1g
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134268 10 g 10g
30138306 2.5 g 2.50g
17904805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134268 Supplier Toronto Research Chemicals Supplier No. TRCM22770010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1120-25-8
Molecular Formula C17 H32 O2
Molecular Weight (g/mol) 268.43
Synonym 9-Hexadecenoic acid, methyl ester, (Z)- (8CI), Palmitoleic acid methyl ester (6CI), (Z)-9-Hexadecenoic acid, methyl ester, Methyl (9Z)-hexadecenoate, Methyl (Z)-9-hexadecenoate, Methyl Z-9-hexadecanoic acid, Methyl cis-9-hexadecenoate, Methyl cis-hexadec-9-enoate, Methyl palmitoleate, Methyl palmitoleinate
IUPAC Name methyl (Z)-hexadec-9-enoate
SMILES CCCCCC\C=C/CCCCCCCC(=O)OC
Specifications
Quantity 10 g
InChI Formula InChI=1S/C17H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h8-9H,3-7,10-16H2,1-2H3/b9-8-
Formula Weight 268.2402
Percent Purity >95%
Chemical Name or Material Methyl Palmitoleate
Grade GC