missing translation for 'onlineSavingsMsg'
Learn More

Methyl-Lysine Antibody, Novus Biologicals™

Product Code. 18208275 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18208275 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18208275 Supplier Novus Biologicals Supplier No. NBP178104
  • ONLINE EXCLUSIVE PRICE
    685.00 EUR
    647.85 EUR / 0.1mg
    Save 37.15 EUR
    5% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Methyl-Lysine Polyclonal specifically detects Methyl-Lysine in Non-species specific samples. It is validated for ELISA, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methyl-Lysine
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:1000-1:20000, Immunoprecipitation
Formulation 0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0, 50% (v/v) Glycerol with 0.01% Sodium Azide
Gene Alias Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine
Host Species Rabbit
Immunogen This affinity-purified Methyl-Lysine Antibody was prepared from whole rabbit serum produced by repeated immunizations with Methyl-Lysine conjugated KLH.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Test Specificity Reactivity is specific for Methylated Lysine residues (mono, di).
Target Species Non-species specific
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.