Learn More
Methyl hydrogen glutarate, 95%
Product Code. 11377276
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11377276
Brand: Thermo Scientific Alfa Aesar L09196.18
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1501-27-5 | C6H10O4 | 146.142 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1501-27-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004409
InChI Key IBMRTYCHDPMBFN-UHFFFAOYSA-N
Synonym monomethyl glutarate, methyl hydrogen glutarate, mono-methyl glutarate, glutaric acid monomethyl ester, monomethyl glutaric acid, 4-methoxycarbonylbutanoic acid, glutaric acid, monomethyl ester, pentanedioic acid, monomethyl ester, 4-methoxycarbonyl butyric acid, glutaric acid methyl half ester
PubChem CID 73917
ChEBI CHEBI:86396
IUPAC Name 5-methoxy-5-oxopentanoic acid
SMILES COC(=O)CCCC(=O)O
Specifications
Boiling Point 150°C to 152°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 1762381
Formula Weight 146.14
Percent Purity 95%
Chemical Name or Material Methyl hydrogen glutarate