CAS RN 1501-27-5
CAS RN 1501-27-5
IUPAC Name:
5-methoxy-5-oxopentanoic acid
Synonyms:
glutaric acid monomethyl ester glutaric acid methyl half ester methyl glutarate,mono
Molecular Weight (g/mol):
146.14
Molecular Formula:
C6H10O4
InChi Key:
IBMRTYCHDPMBFN-UHFFFAOYSA-N
SMILES:
COC(=O)CCCC(O)=O
1
–
1
of
1
results
1
Methyl hydrogen glutarate, 95%
CAS: 1501-27-5 Molecular Formula: C6H10O4 Molecular Weight (g/mol): 146.142 MDL Number: MFCD00004409 InChI Key: IBMRTYCHDPMBFN-UHFFFAOYSA-N Synonym: monomethyl glutarate,methyl hydrogen glutarate,mono-methyl glutarate,glutaric acid monomethyl ester,monomethyl glutaric acid,4-methoxycarbonylbutanoic acid,glutaric acid, monomethyl ester,pentanedioic acid, monomethyl ester,4-methoxycarbonyl butyric acid,glutaric acid methyl half ester PubChem CID: 73917 ChEBI: CHEBI:86396 IUPAC Name: 5-methoxy-5-oxopentanoic acid SMILES: COC(=O)CCCC(=O)O