Learn More
Methyl 4-chlorocinnamate, 99%
Product Code. 11408936
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408936 5 g 5g
11498926 1 g 1g
11418936 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11408936 Supplier Thermo Scientific Alfa Aesar Supplier No. A17071.06
 Restricted Item
In Stock
Add to basket
CAS: 7560-44-3 | C10H9ClO2 | 196.63 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7560-44-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00075790
InChI Key IIBXQGYKZKOORG-QPJJXVBHSA-N
Synonym methyl 4-chlorocinnamate, methyl p-chlorocinnamate, methyl 3-4-chlorophenyl acrylate, 4-chlorocinnamic acid methyl ester, cinnamic acid, p-chloro-, methyl ester, 2-propenoic acid, 3-4-chlorophenyl-, methyl ester, methyl e-3-4-chlorophenyl prop-2-enoate, e-methyl 3-4-chlorophenyl acrylate, methyl 2e-3-4-chlorophenyl prop-2-enoate, methyl 3-4-chlorophenyl prop-2-enoate
PubChem CID 5314314
IUPAC Name methyl (E)-3-(4-chlorophenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=C1)Cl
Specifications
Melting Point 74°C to 77°C
Quantity 5 g
Beilstein 2045629
Formula Weight 196.64
Percent Purity 99%
Chemical Name or Material Methyl 4-chlorocinnamate