accessibility menu, dialog, popup

UserName

CH3OOCl(E)

CAS RN 7560-44-3

IUPAC Name: methyl (2E)-3-(4-chlorophenyl)prop-2-enoate
Synonyms: methyl (2E)-3-(4-chlorophenyl)prop-2-enoate
Molecular Weight (g/mol): 196.63
Molecular Formula: C10H9ClO2
InChi Key: IIBXQGYKZKOORG-QPJJXVBHSA-N
SMILES: COC(=O)\C=C\C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results