missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxybutyrate, TRC

Product Code. 30484234
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484234 50 mg 50mg
30468228 100 mg 100mg
30465310 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484234 Supplier Toronto Research Chemicals Supplier No. TRCM31258350MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1487-49-6
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
Synonym Butyric acid, 3-hydroxy-, methyl ester (6CI,7CI,8CI), (±)-Methyl 3-hydroxybutanoate, 3-Hydroxybutyric acid methyl ester, Methyl (RS)-3-hydroxybutyrate, Methyl 3-hydroxybutanoate, Methyl beta-hydroxybutyrate
IUPAC Name methyl 3-hydroxybutanoate
SMILES COC(=O)CC(C)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H10O3/c1-4(6)3-5(7)8-2/h4,6H,3H2,1-2H3
Formula Weight 118.063
Chemical Name or Material Methyl 3-Hydroxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.