CAS RN 1487-49-6
CAS RN 1487-49-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 3-Hydroxybutyrate, TRC
CAS: 1487-49-6 Molecular Formula: C5H10O3 Molecular Weight (g/mol): 118.13 Synonym: Butyric acid, 3-hydroxy-, methyl ester (6CI,7CI,8CI),(±)-Methyl 3-hydroxybutanoate,3-Hydroxybutyric acid methyl ester,Methyl (RS)-3-hydroxybutyrate,Methyl 3-hydroxybutanoate,Methyl beta-hydroxybutyrate IUPAC Name: methyl 3-hydroxybutanoate SMILES: COC(=O)CC(C)O