missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Methyl 2-fluoro-6-methoxybenzoate, 98%
Description
Applications
Methyl 2-Fluoro-6-methoxybenzoate is used in the synthetic preparation of thiazinotrienomycin E which is an ansamycin macrolactam antibiotic. Useful intermediates for the synthesis of thiazolones and 2,4,5-trisubstituted thiazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Methyl 2-Fluoro-6-methoxybenzoate is used in the synthetic preparation of thiazinotrienomycin E which is an ansamycin macrolactam antibiotic. Useful intermediates for the synthesis of thiazolones and 2,4,5-trisubstituted thiazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Refractive Index | 1.501 |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 184.17 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 2-fluoro-6-methoxybenzoate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?