accessibility menu, dialog, popup

UserName

H3COFOOCH3

CAS RN 178747-79-0

IUPAC Name: methyl 2-fluoro-6-methoxybenzoate
Synonyms: methyl 2-fluoro-6-methoxybenzoate
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: WOKPPDUGASITJX-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results