missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-ethylacetoacetate, 97%

Product Code. 11499307
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499307 100 g 100g
11489307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499307 Supplier Alfa Aesar™ Supplier No. B21368.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51756-08-2
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD01632136
InChI Key YXLVLOWNJCOOAU-UHFFFAOYSA-N
Synonym methyl 2-ethylacetoacetate, butanoic acid, 2-ethyl-3-oxo-, methyl ester, methyl-2-ethylacetoacetate, 2-ethyl-3-oxobutanoic acid, methyl ester, methyl ethylacetoacetate, methyl acetylbutyrate, methyl 2-acetylbutyrate, acmc-1b06q, methyl 2-ethyl-3-oxobutyrate, methyl 2-ethyl-3-oxo-butyrate
PubChem CID 103534
IUPAC Name methyl 2-ethyl-3-oxobutanoate
SMILES CCC(C(=O)C)C(=O)OC

Specifications

Density 1.01
Boiling Point 189°C to 191°C
Refractive Index 1.423
Quantity 100 g
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Methyl 2-ethylacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.