CAS RN 51756-08-2
IUPAC Name:
methyl 2-ethyl-3-oxobutanoate
Synonyms:
methyl ethylacetoacetate
Molecular Weight (g/mol):
144.17
Molecular Formula:
C7H12O3
InChi Key:
YXLVLOWNJCOOAU-UHFFFAOYNA-N
SMILES:
CCC(C(C)=O)C(=O)OC
1
–
1
of
1
results
1
Alfa Aesar™ Methyl 2-ethylacetoacetate, 97%
CAS: 51756-08-2 Molecular Formula: C7H12O3 Molecular Weight (g/mol): 144.17 MDL Number: MFCD01632136 InChI Key: YXLVLOWNJCOOAU-UHFFFAOYSA-N Synonym: methyl 2-ethylacetoacetate,butanoic acid, 2-ethyl-3-oxo-, methyl ester,methyl-2-ethylacetoacetate,2-ethyl-3-oxobutanoic acid, methyl ester,methyl ethylacetoacetate,methyl acetylbutyrate,methyl 2-acetylbutyrate,acmc-1b06q,methyl 2-ethyl-3-oxobutyrate,methyl 2-ethyl-3-oxo-butyrate PubChem CID: 103534 IUPAC Name: methyl 2-ethyl-3-oxobutanoate SMILES: CCC(C(=O)C)C(=O)OC