missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-diphenylphosphino-1-naphthoate, 98%, Thermo Scientific™

Product Code. 15464007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464007 5 g 5g
15454007 1 g 1g
15474007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15464007 Supplier Thermo Scientific Alfa Aesar Supplier No. H30928.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178176-78-8
Molecular Formula C24H19O2P
Molecular Weight (g/mol) 370.39
MDL Number MFCD12546191
InChI Key QNDSZNKYBYBYAE-UHFFFAOYSA-N
Synonym methyl 2-diphenylphosphino-1-naphthoate, 1-carbomethoxy-2-diphenylphosphino naphthalene, 2-diphenylphosphino-1-naphthoic acid methyl ester, methyl 2-diphenylphosphanyl naphthalene-1-carboxylate
PubChem CID 11760420
IUPAC Name methyl 2-diphenylphosphanylnaphthalene-1-carboxylate
SMILES COC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 107°C to 108°C
Quantity 5 g
Formula Weight 370.39
Percent Purity 98%
Chemical Name or Material Methyl 2-diphenylphosphino-1-naphthoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.