accessibility menu, dialog, popup

UserName

CH3OOP

CAS RN 178176-78-8

IUPAC Name: methyl 2-(diphenylphosphanyl)naphthalene-1-carboxylate
Synonyms: methyl 2-(diphenylphosphanyl)naphthalene-1-carboxylate
Molecular Weight (g/mol): 370.39
Molecular Formula: C24H19O2P
InChi Key: QNDSZNKYBYBYAE-UHFFFAOYSA-N
SMILES: COC(=O)C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results