missing translation for 'onlineSavingsMsg'
Learn More

MBD1 Antibody, Novus Biologicals™

Product Code. 18214903 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214903 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214903 Supplier Novus Biologicals Supplier No. NBP259293
  • ONLINE EXCLUSIVE PRICE
    500.00 EUR
    472.50 EUR / 50µg
    Save 27.50 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MBD1 Polyclonal specifically detects MBD1 in Human, Mouse samples. It is validated for ChIP Assay, ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD1
Applications ChIP Assay, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:1000, Chromatin Immunoprecipitation (ChIP) 1.5 ug
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Gene Symbols MBD1
Host Species Rabbit
Immunogen The exact immunogen is propietary information.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human, Mouse
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.