missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysozyme, MP Biomedicals™

Product Code. 11374019 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374019 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11374019 Supplier MP Biomedicals™ Supplier No. 02100831.5
  • ONLINE EXCLUSIVE PRICE
    46.30 EUR
    44.10 EUR / 500mg
    Save 2.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for lysing E. coli and Streptomycetes for extraction purposes. Lysozyme, MP Biomedicals preferentially hydrolyzes the β-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine. Enzyme activity: ≥20,000u/mg solid.

An enzyme used in a variety of biochemical applications.

  • For lysing E. coli and Streptomycetes for extraction purposes, such as extracting group specific antigen
  • May act as a germinative agent of bacterial spores
  • In humans, may be the mediator in the anti-tumor function of macrophages which secrete the enzyme
  • There is evidence that cartilage lysozyme has a role in cartilage calcification
  • The enzyme breaks down the cell walls of bacteria
  • Used to prepare spheroplasts

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin.

  • These links occur in the mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus.
  • Gram-positive cells are quite susceptible to this hydrolysis, as their cell walls have a high proportion of peptidoglycan.
  • Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.
  • The enzyme is active over a broad pH range (6.0–9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02–0.100M) than at pH 9.2 (0.01–0.06M).

Specifications

Concentration Greater than or equal to 20,000 U/mg
Color White
Description Lysozyme
CAS 9001-63-2
Formula Weight 2235.597 g/mol
Synonym Muramidase; Mucopeptide N-acetylmuramoylhydrolase
Content And Storage Store at -20°C
Donor Source Chicken Egg White
Form Powder
For Use With (Application) Cell Lysis
Quantity 500 mg
Show More Show Less
compliance-icons
Product Identifier
  • Lysozyme
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: lysozyme
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.