Learn More
Lithium Carbonate, ≥99%, (ACS Reagent Grade), MP Biomedicals
Product Code. 11482862 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482862 500 g 500g
11472862 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11482862 Supplier MP Biomedicals Supplier No. 0215253790
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity ACS reagent grade lithium carbonate for laboratory applications.
  • Grade: ACS Reagent
  • Assay: ≥99%
  • Molecular Formula: CLi2O3
  • Molecular Weight: 73.89 g/mol
  • CAS Number: 554-13-2
  • Storage: Room temperature
Chemical Identifiers
CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O
Specifications
Melting Point 723°C
Color White
pH 11
Physical Form Powder
Quantity 500 g
Loss on Drying max. 1%
Odor Odorless
Packaging Poly Bottle
Sensitivity Hygroscopic
Appearance White Powder
Specific Gravity 2.11
Assay Percent Range ≥99%
Decomposition Information Carbon Oxides; Lithium Oxides
Density 2.11 g/cm³
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3586314
Merck Index 15, 5506
Solubility Information Soluble in water
Formula Weight 73.9
Percent Purity ≥99%
Grade Meets ACS reagent chemical requirements
Manufacturing Origin USA
Chemical Name or Material Lithium Carbonate