missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate, ≥99%, (ACS Reagent Grade), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | ≥99% |
| Formula Weight | 73.9 |
| Grade | ACS Reagent |
| Chemical Name or Material | Lithium Carbonate |
Safety and Handling
Product Identifier
- Lithium Carbonate, ACS
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : lithium carbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction