missing translation for 'onlineSavingsMsg'
Learn More

Leptin R Antibody (RM0085-9C29), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18214735 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214735 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214735 Supplier Novus Biologicals Supplier No. NBP121725AF532
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody

Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leptin R
Applications Western Blot
Classification Monoclonal
Clone RM0085-9C29
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Rat
Immunogen Mouse recombinant EC-domain of LeptinR
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3953.0
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.