missing translation for 'onlineSavingsMsg'
Learn More

Leptin/OB Mouse anti-Human, Clone: 7.00E+10, Novus Biologicals™

Product Code. 18214754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214754 50 μL 50µL
18031204 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18214754 Supplier Novus Biologicals Supplier No. NBP25949750UL
  • ONLINE EXCLUSIVE PRICE
    315.00 EUR
    297.15 EUR / 50µL
    Save 17.85 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leptin/OB
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 70000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Gene Symbols LEP
Host Species Mouse
Immunogen Recombinant human Leptin (22-167aa) purified from E. coli.
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3952
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.