missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MP Biomedicals™

Product Code. 11438171 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
Unit Size:
100g
250g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438171 25 g 25g
11448171 100 g 100g
11458171 250 g 250g
11468171 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11438171 Supplier MP Biomedicals Supplier No. 0215153925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an emulsifier and solubilizer for drugs in aqueous solution and as a vehicle for liposomes. Lecithin, MP Biomedicals is a major structural phospholipid in the brain, comprising ∼15% of total lipid and localized to gray matter. It is a purified

Lecithin (aka phosphatidylcholine) is the major structural component of cellular membranes and serves as a reservoir for several lipid messengers. It is the source of the bioactive lipids: lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, platelet activating factor and arachidonic acid.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 25 g
Percent Purity ≥90%
Grade Water Dispersible
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.