CAS RN 8002-43-5
Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 750 MDL Number: MFCD00082428 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Thermo Scientific Chemicals Lecithin, Refined
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00147406 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, 90%, soybean
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00147406 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, ∽60%, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Lecithin (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Melford Lecithin
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Lecithin (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Lecithin, MedChemExpress
MedChemExpress Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.