missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MP Biomedicals™

Product Code. 11490242 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
5 g
Unit Size:
100mg
250mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490242 100 mg 100mg
11400252 250 mg 250mg
11410252 500 mg 500mg
11430252 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11490242 Supplier MP Biomedicals Supplier No. 02160059.1
  • ONLINE EXCLUSIVE PRICE
    110.00 EUR
    103.95 EUR / 100mg
    Save 6.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serves as a reservoir for several lipid messengers. Lecithin, MP Biomedicals is the major membrane phospholipid in eukaryotic cells. It is the major structural phospholipid in the brain, comprising approx. 15% of the total lipid and is primarily localized

Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers. It is the source of the bioactive lipids lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, plateletactivating factor, and arachidonic acid.

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 100 mg
Solubility Information Soluble in fat, paraffin, ether, ethanol, chlorinated hydrocarbons(100mg/mL, CHCl3). Dispersible in water.
Percent Purity ≥96%
Physical Form Fluffy Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.