Learn More
Lauric Acid, Sigma-Aldrich™
Product Code. 15819190
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819190 1 kg 1kg
15809190 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15819190 Supplier Sigma Aldrich Supplier No. 8053331000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143-07-7
Specifications
Melting Point 43°C to 45°C
Density 0.883 g/cm3 at 50°C
Boiling Point 299°C/1013 hPa
Molecular Formula CH3(CH2)10COOH
MDL Number MFCD00002736
Quantity 1 kg
Synonym Lauric acid, Dodecanoic acid, Lauric acid
Molecular Weight (g/mol) 200.32
Grade Research