accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 143-07-7

IUPAC Name: dodecanoic acid
Synonyms: lauric acid laurinsaeure L556_aldrich N-dodecanoic acid prifrac 2920 laurostearic acid dodecanoate ABL DAO univol U-314
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: POULHZVOKOAJMA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (11)
  • (7)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (6)
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (5)
  • (3)
Physical Form 
  • (3)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (5)
  • (6)
  • (3)
  • (4)
  • (1)
  • (11)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)