missing translation for 'onlineSavingsMsg'
Learn More

LANA2 Antibody (CM-A807), DyLight 550, Novus Biologicals™

Product Code. 18215010 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215010 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215010 Supplier Novus Biologicals Supplier No. NB200167R
  • ONLINE EXCLUSIVE PRICE
    509.00 EUR
    480.90 EUR / 0.1mL
    Save 28.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LANA2 Monoclonal specifically detects LANA2 in Human, Mouse, Virus samples. It is validated for ELISA, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LANA2
Applications ELISA, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CM-A807
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated
Gene Alias LANA 2, LANA-2, Latency-associated nuclear antigen 2, vIRF 3, vIRF3, vIRF-3
Host Species Mouse
Immunogen Recombinant LANA2 fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Chromatin Research, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This is specific for KSHV LANA2 (residues 143-310 epitope recognition).
Target Species Human, Mouse, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.