missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Antibody (LLC/1738) - Azide and BSA Free, Novus Biologicals™

Product Code. 18214640 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214640 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214640 Supplier Novus Biologicals Supplier No. NBP254420100UG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgL Monoclonal specifically detects IgL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgL
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LLC/1738
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias ig lambda-6 chain C region, IGL, IGLC6, immunoglobulin lambda gene cluster, immunoglobulin lambda locus, Lambda Light Chain, MGC88804
Gene Symbols IGL
Host Species Mouse
Immunogen Purified human full-length lambda light chain
Molecular Weight of Antigen 22.5 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3535
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.