Learn More
L-Quebrachitol, TRC
Product Code. 30418996
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418996 100 mg -
30410142 500 mg 500mg
30413743 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30418996 Supplier Toronto Research Chemicals Supplier No. TRCQ990090100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 642-38-6
Molecular Formula C7 H14 O6
Molecular Weight (g/mol) 194.18
Synonym 2-O-methyl-L-chiro-inositol, Brahol, Quebrachit
IUPAC Name (1R,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol
SMILES CO[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C7H14O6/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h2-12H,1H3/t2-,3-,4-,5+,6+,7-/m0/s1
Formula Weight 194.079
Chemical Name or Material (-)-Quebrachitol