accessibility menu, dialog, popup

UserName

H3CO(R)(R)OH(S)OH(S)OH(S)HO(R)HO

CAS RN 642-38-6

IUPAC Name: (1R,2S,3S,4S,5R,6R)-6-methoxycyclohexane-1,2,3,4,5-pentol
Synonyms: (-)-quebrachitol quebrachitol
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: DSCFFEYYQKSRSV-FIZWYUIZSA-N
SMILES: CO[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)