missing translation for 'onlineSavingsMsg'
Learn More

δ-Ketoendrin, TRC

Product Code. 18093330
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093330 100 mg 100mg
18013240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18093330 Supplier Toronto Research Chemicals Supplier No. TRCK188700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53494-70-5
Molecular Formula C12 H8 Cl6 O
Molecular Weight (g/mol) 380.91
Synonym 2,5,7-Metheno-3H-cyclopenta[a]pentalen-3-one, 3b,4,5,6,6,6a-hexachlorodecahydro-, (2R,3aR,3bS,4R,5R,6aS,7S,7aS,8R)-rel-, 2,5,7-Metheno-3H-cyclopenta[a]pentalen-3-one, 3b,4,5,6,6,6a-hexachlorodecahydro-, (2α,3aβ,3bβ,4β,5β,6aβ,7α,7aβ,8R*)-, rel-(2R,3aR,3bS,4R,5R,6aS,7S,7aS,8R)-3b,4,5,6,6,6a-Hexachlorodecahydro-2,5,7-metheno-3H-cyclopenta[a]pentalen-3-one, Endrin ketone, SD 2614, δ-Ketoendrin
IUPAC Name rel-(2R,3aR,3bS,4R,5R,6aS,7S,7aS,8R)-3b,4,5,6,6,6a-Hexachlorodecahydro-2,5,7-metheno-3H-cyclopenta[a]pentalen-3-one
SMILES Cl[C@H]1[C@]2(Cl)[C@@H]3[C@H]4C[C@H]5[C@@H]3[C@@](Cl)(C2(Cl)Cl)[C@]1(Cl)[C@H]5C4=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H8Cl6O/c13-8-9(14)6-2-1-3(7(6)19)4-5(2)11(9,16)12(17,18)10(4,8)15/h2-6,8H,1H2/t2-,3+,4+,5-,6+,8+,9-,10+,11-/m0/s1
Formula Weight 377.8706
Percent Purity >95%
Chemical Name or Material Endrin-ketone
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.