accessibility menu, dialog, popup

UserName

ClClClClClClO

CAS RN 53494-70-5

IUPAC Name: 1,2,2,3,10,11-hexachloropentacyclo[5.4.1.0³,¹⁰.0⁴,¹².0⁵,⁹]dodecan-8-one
Synonyms: endrin ketone
Molecular Weight (g/mol): 380.9
Molecular Formula: C12H8Cl6O
InChi Key: IZHZFAQWVKBTSL-UHFFFAOYNA-N
SMILES: ClC1C2(Cl)C3C4CC5C3C(Cl)(C2(Cl)Cl)C1(Cl)C5C4=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
Concentration 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)