missing translation for 'onlineSavingsMsg'
Learn More

Isosulfamethoxazole, TRC

Product Code. 30008755
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008755 10 mg 10mg
30004882 50 mg 50mg
30004067 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008755 Supplier Toronto Research Chemicals Supplier No. TRCI90223010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17103-52-5
Molecular Formula C10 H11 N3 O3 S
Molecular Weight (g/mol) 253.28
Synonym Sulfamethoxazole EP impurity F
IUPAC Name 4-amino-N-(3-methyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES Cc1cc(NS(=O)(=O)c2ccc(N)cc2)on1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H11N3O3S/c1-7-6-10(16-12-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6,13H,11H2,1H3
Formula Weight 253.0521
Percent Purity >95%
Chemical Name or Material 4-amino-N-(3-methylisoxazol-5-yl)benzenesulfonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.