CAS RN 17103-52-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isosulfamethoxazole, TRC
CAS: 17103-52-5 Molecular Formula: C10 H11 N3 O3 S Molecular Weight (g/mol): 253.28 Synonym: Sulfamethoxazole EP impurity F IUPAC Name: 4-amino-N-(3-methyl-1,2-oxazol-5-yl)benzenesulfonamide SMILES: Cc1cc(NS(=O)(=O)c2ccc(N)cc2)on1