Learn More
Isopropyl methanesulfonate, 99%
Product Code. p-152061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15246483 5 g Glass Bottle 5g
10677191 25 g Glass Bottle 25g
15204734 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15246483 Supplier Thermo Scientific Acros Supplier No. 412840050
Add to basket
CAS: 926-06-7 | C4H10O3S | 138.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 926-06-7
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00047802
InChI Key SWWHCQCMVCPLEQ-UHFFFAOYSA-N
Synonym isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26b
PubChem CID 13551
IUPAC Name propan-2-yl methanesulfonate
SMILES CC(C)OS(=O)(=O)C
Specifications
Color Colorless
Density 1.1450g/mL
Boiling Point 82.0°C (6.0 mmHg)
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3SO3CH(CH3)2
Refractive Index 1.4180 to 1.4210
Quantity 5 g
UN Number 1760
Beilstein 04,IV,13
Specific Gravity 1.145
Formula Weight 138.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl Methanesulfonate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropyl methanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.