accessibility menu, dialog, popup

UserName

CAS RN 926-06-7

IUPAC Name: propan-2-yl methanesulfonate
Synonyms: isopropylmethanesulfonate
Molecular Weight (g/mol): 138.18
Molecular Formula: C4H10O3S
InChi Key: SWWHCQCMVCPLEQ-UHFFFAOYSA-N
SMILES: CC(C)OS(C)(=O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results