missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Benzenesulfonate, TRC

Product Code. 17940425
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940425 100 mg 100mg
17903385 25 mg 25mg
17920415 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17940425 Supplier Toronto Research Chemicals Supplier No. TRCI824350100MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6214-18-2
Molecular Formula C9 H12 O3 S
Molecular Weight (g/mol) 200.25
Synonym Benzenesulfonic Acid Isopropyl Ester, Benzenesulfonic Acid 1-Methylethyl Ester; Isopropyl Besylate;
IUPAC Name propan-2-yl benzenesulfonate
SMILES CC(C)OS(=O)(=O)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H12O3S/c1-8(2)12-13(10,11)9-6-4-3-5-7-9/h3-8H,1-2H3
Formula Weight 200.0507
Percent Purity >95%
Chemical Name or Material Isopropyl Benzenesulfonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.