accessibility menu, dialog, popup

UserName

H3CCH3OSOO

CAS RN 6214-18-2

IUPAC Name: 2-methylpropyl benzenesulfonate
Synonyms: 2-methylpropyl benzenesulfonate
Molecular Weight (g/mol): 214.28
Molecular Formula: C10H14O3S
InChi Key: NFRRFEFZKGNYKS-UHFFFAOYSA-N
SMILES: CC(C)COS(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results