Learn More
Isopropyl Benzenesulfonate, TRC
Product Code. 17903385
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903385 25 mg 25mg
17920415 10 mg 10mg
17940425 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17903385 Supplier Toronto Research Chemicals Supplier No. TRCI82435025MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6214-18-2
Molecular Formula C9 H12 O3 S
Molecular Weight (g/mol) 200.25
Synonym Benzenesulfonic Acid Isopropyl Ester, Benzenesulfonic Acid 1-Methylethyl Ester; Isopropyl Besylate;
IUPAC Name propan-2-yl benzenesulfonate
SMILES CC(C)OS(=O)(=O)c1ccccc1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C9H12O3S/c1-8(2)12-13(10,11)9-6-4-3-5-7-9/h3-8H,1-2H3
Formula Weight 200.0507
Percent Purity >95%
Chemical Name or Material Isopropyl Benzenesulfonate
Grade HPLC