Learn More
Isonicoteine, TRC
Product Code. 30132720
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132720 25 mg 25mg
30009470 5 mg 5mg
30011494 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132720 Supplier Toronto Research Chemicals Supplier No. TRCI82167525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 581-50-0
Molecular Formula C10 H8 N2
Molecular Weight (g/mol) 156.18
IUPAC Name 2-pyridin-3-ylpyridine
SMILES c1ccc(nc1)c2cccnc2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10H8N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-8H
Formula Weight 156.0687
Percent Purity >95%
Chemical Name or Material Isonicoteine
Grade GC