CAS RN 581-50-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isonicoteine, TRC
CAS: 581-50-0 Molecular Formula: C10 H8 N2 Molecular Weight (g/mol): 156.18 IUPAC Name: 2-pyridin-3-ylpyridine SMILES: c1ccc(nc1)c2cccnc2