Learn More
Isodesmosine, ≥99%, MP Biomedicals™
Product Code. 11440452 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440452 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11440452 Supplier MP Biomedicals Supplier No. 0219137901
  • ONLINE EXCLUSIVE PRICE
    182.00 EUR
    173.25 EUR / 1mg
    Save 8.75 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Molecular biology
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 991-01-5
Molecular Formula C24H40N5O8
Molecular Weight (g/mol) 526.61
InChI Key RGXCTRIQQODGIZ-UHFFFAOYNA-O
Synonym isodesmosine, 2-amino-6-3,5-bis 3-amino-4-hydroxy-4-oxobutyl-2-4-amino-5-hydroxy-5-oxopentyl pyridin-1-ium-1-yl hexanoic acid, iso-desmosin, 6-2-4-amino-4-carboxybutyl-3,5-bis 3-amino-3-carboxypropyl pyridinium-1-yl norleucine, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl-pyridinium, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl pyridinium, 3,5-bis 3-amino-3-carboxypropyl-2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl pyridin-1-ium, pyridinium, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl
PubChem CID 13811
ChEBI CHEBI:64366
Specifications
Color Yellow
Assay Percent Range ≥99%
Quantity 1 mg
Formula Weight 526
Physical Form Solid
Chemical Name or Material Isodesmosine
compliance-icons
Product Identifier
  • isodesmosine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: isodesmosine